Assay Details
| Gene Symbol: CBFA2T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011689.1 | NP_001011689.1 | 5 - 6 | 729 | 62 | |
| GenBank mRNA | AY714076.1 | - | 5 - 6 | 729 | 62 | |
More Information
Set Membership: |
|

CBFA2T2
Genomic Map