Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011692.1 | NP_001011692.1 | 2 - 3 | 202 | 67 | |
| GenBank mRNA | AY735998.1 | - | 2 - 3 | 202 | 67 | |
| BX934317.2 | - | 2 - 3 | 203 | 67 | ||
More Information
Set Membership: |
|

APOD
Genomic Map