Assay Details
| Gene Symbol: PRLHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012295.2 | NP_001012295.1 | 1 | 750 | 84 | |
| XM_015288694.2 | XP_015144180.1 | 4 | 2590 | 84 | ||
| XM_015288696.2 | XP_015144182.1 | 4 | 1595 | 84 | ||
| XM_025151643.1 | XP_025007411.1 | 3 | 1541 | 84 | ||
| GenBank mRNA | AY847003.1 | - | 1 | 751 | 84 | |
More Information
Set Membership: |
|

PRLHR
Genomic Map