Assay Details
| Gene Symbol: SPG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012527.2 | NP_001012545.1 | 2 - 3 | 335 | 71 | |
| GenBank mRNA | AJ851745.1 | - | 2 - 3 | 337 | 71 | |
More Information
Set Membership: |
|

SPG7
Genomic Map