Assay Details
| Gene Symbol: PDPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012529.1 | NP_001012547.1 | 12 - 13 | 1512 | 64 | |
| GenBank mRNA | AJ851556.1 | - | 12 - 13 | 1512 | 64 | |
More Information
Set Membership: |
|

PDPK1
Genomic Map