Assay Details
| Gene Symbol: MAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012555.3 | NP_001012573.2 | 2 - 3 | 374 | 109 | |
| GenBank mRNA | AJ448285.1 | - | 2 - 3 | 376 | 109 | |
| AJ851580.1 | - | 2 - 3 | 366 | 109 | ||
More Information
Set Membership: |
|

MAL2
Genomic Map