Assay Details
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012560.1 | NP_001012578.1 | 4 - 5 | 537 | 70 | |
XM_015275865.2 | XP_015131351.1 | 4 - 5 | 553 | 70 | ||
XM_015275868.2 | XP_015131354.1 | 4 - 5 | 553 | 70 | ||
XM_015275869.2 | XP_015131355.1 | 4 - 5 | 553 | 70 | ||
XM_015275870.2 | XP_015131356.1 | 5 - 6 | 697 | 70 | ||
XM_015275871.2 | XP_015131357.1 | 4 - 5 | 538 | 70 | ||
XM_025147252.1 | XP_025003020.1 | 4 - 5 | 553 | 70 | ||
XM_025147253.1 | XP_025003021.1 | 5 - 6 | 698 | 70 | ||
XM_025147254.1 | XP_025003022.1 | 4 - 5 | 441 | 70 | ||
XM_025147255.1 | XP_025003023.1 | 3 - 4 | 379 | 70 | ||
XM_025147256.1 | XP_025003024.1 | 4 - 5 | 553 | 70 | ||
GenBank mRNA | AJ851478.1 | - | 4 - 5 | 537 | 70 | |
CR389346.1 | - | 2 - 3 | 139 | 70 | ||
CR406494.1 | - | 5 - 6 | 646 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
