Assay Details
| Gene Symbol: SH3BGRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012574.1 | NP_001012592.1 | 2 - 3 | 254 | 71 | |
| GenBank mRNA | AJ851721.1 | - | 2 - 3 | 254 | 71 | |
More Information
Set Membership: |
|

SH3BGRL
Genomic Map