Assay Details
Gene Symbol: HNRNPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012592.1 | NP_001012610.1 | 5 - 6 | 735 | 69 | |
XM_015278681.2 | XP_015134167.1 | 5 - 6 | 690 | 69 | ||
XM_015278682.2 | XP_015134168.1 | 5 - 6 | 688 | 69 | ||
XM_015278683.2 | XP_015134169.1 | 5 - 6 | 733 | 69 | ||
XM_025151554.1 | XP_025007322.1 | 5 - 6 | 614 | 69 | ||
XM_025151555.1 | XP_025007323.1 | 5 - 6 | 646 | 69 | ||
XM_025151556.1 | XP_025007324.1 | 5 - 6 | 666 | 69 | ||
XM_025151557.1 | XP_025007325.1 | 5 - 6 | 639 | 69 | ||
XM_025151558.1 | XP_025007326.1 | 5 - 6 | 648 | 69 | ||
XM_025151560.1 | XP_025007328.1 | 4 - 5 | 489 | 69 | ||
XM_025151561.1 | XP_025007329.1 | 4 - 5 | 482 | 69 | ||
XM_025151562.1 | XP_025007330.1 | 4 - 5 | 604 | 69 | ||
GenBank mRNA | AJ851718.1 | - | 5 - 6 | 735 | 69 | |
CR352387.1 | - | 1 - 2 | 33 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
