Assay Details
| Gene Symbol: GORASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012594.2 | NP_001012612.1 | 8 - 9 | 976 | 60 | |
| XM_015289644.2 | XP_015145130.1 | 8 - 9 | 950 | 60 | ||
| GenBank mRNA | AJ851797.1 | - | 8 - 9 | 978 | 60 | |
More Information
Set Membership: |
|

GORASP2
Genomic Map