Assay Details
| Gene Symbol: SEC62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012602.1 | NP_001012620.1 | 7 - 8 | 740 | 75 | |
| GenBank mRNA | AJ851749.1 | - | 7 - 8 | 740 | 75 | |
More Information
Set Membership: |
|

SEC62
Genomic Map