Assay Details
| Gene Symbol: TIPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012690.1 | NP_001012708.1 | 2 - 3 | 285 | 69 | |
| XM_015292095.2 | XP_015147581.1 | 2 - 3 | 610 | 69 | ||
| GenBank mRNA | AJ851484.1 | - | 2 - 3 | 285 | 69 | |
More Information
Set Membership: |
|

TIPIN
Genomic Map