Assay Details
| Gene Symbol: LOC112530170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142834.1 | XP_024998602.1 | 2 - 3 | 350 | 76 | |
| Gene Symbol: SNRNP200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ851437.1 | - | 5998 | 76 | ||
More Information
Set Membership: |
|

LOC112530170
Genomic Map