Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012760.1 | NP_001012778.1 | 1 - 2 | 2832 | 72 | |
| XM_015275303.2 | XP_015130789.1 | 2 - 3 | 3315 | 72 | ||
| GenBank mRNA | D82364.1 | - | 1 - 2 | 2832 | 72 | |
More Information
Set Membership: |
|

TSC22D1
Genomic Map