Assay Details
| Gene Symbol: SCPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012803.1 | NP_001012821.1 | 1 - 2 | 104 | 83 | |
| XM_015279876.2 | XP_015135362.1 | 1 - 2 | 129 | 83 | ||
| GenBank mRNA | AJ851536.1 | - | 1 - 2 | 104 | 83 | |
More Information
Set Membership: |
|

SCPEP1
Genomic Map