Assay Details
| Gene Symbol: VPS53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012806.1 | NP_001012824.1 | 21 - 22 | 2337 | 59 | |
| GenBank mRNA | AJ719797.1 | - | 21 - 22 | 2337 | 59 | |
More Information
Set Membership: |
|

VPS53
Genomic Map