Assay Details
| Gene Symbol: ACOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012823.1 | NP_001012841.1 | 7 - 8 | 666 | 66 | |
| XM_015302437.2 | XP_015157923.1 | 8 - 9 | 756 | 66 | ||
| XM_015302447.1 | XP_015157933.1 | 7 - 8 | 591 | 66 | ||
| XM_025149635.1 | XP_025005403.1 | 9 - 10 | 639 | 66 | ||
| XM_025149637.1 | XP_025005405.1 | 7 - 8 | 672 | 66 | ||
| XM_025149642.1 | XP_025005410.1 | 6 - 7 | 599 | 66 | ||
| XM_025149643.1 | XP_025005411.1 | 6 - 7 | 1483 | 66 | ||
| XM_025149645.1 | XP_025005413.1 | 6 - 7 | 632 | 66 | ||
| GenBank mRNA | AJ851736.1 | - | 7 - 8 | 666 | 66 | |
More Information
Set Membership: |
|

ACOT9
Genomic Map