Assay Details
| Gene Symbol: SLC25A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012883.1 | NP_001012901.1 | 3 - 4 | 375 | 75 | |
| XM_015278211.2 | XP_015133697.1 | 3 - 4 | 347 | 75 | ||
| GenBank mRNA | AJ720306.1 | - | 3 - 4 | 375 | 75 | |
| CR354223.1 | - | 3 - 4 | 350 | 75 | ||
More Information
Set Membership: |
|

SLC25A14
Genomic Map