Assay Details
| Gene Symbol: CTBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012890.1 | NP_001012908.1 | 6 - 7 | 895 | 73 | |
| XM_025149718.1 | XP_025005486.1 | 6 - 7 | 931 | 73 | ||
| XM_025149719.1 | XP_025005487.1 | 6 - 7 | 931 | 73 | ||
| XM_025149720.1 | XP_025005488.1 | 7 - 8 | 1091 | 73 | ||
| XM_025149721.1 | XP_025005489.1 | 7 - 8 | 1114 | 73 | ||
| XM_025149722.1 | XP_025005490.1 | 6 - 7 | 1041 | 73 | ||
| XM_025149723.1 | XP_025005491.1 | 5 - 6 | 759 | 73 | ||
| GenBank mRNA | AJ720638.1 | - | 6 - 7 | 895 | 73 | |
More Information
Set Membership: |
|

CTBP1
Genomic Map