Assay Details
| Gene Symbol: SLC40A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012913.1 | NP_001012931.1 | 3 - 4 | 497 | 68 | |
| XM_015289163.2 | XP_015144649.1 | 4 - 5 | 594 | 68 | ||
| GenBank mRNA | AJ851647.1 | - | 3 - 4 | 497 | 68 | |
More Information
Set Membership: |
|

SLC40A1
Genomic Map