Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012943.1 | NP_001012961.1 | 3 - 4 | 301 | 69 | |
| XM_015284114.2 | XP_015139600.1 | 3 - 4 | 312 | 69 | ||
| GenBank mRNA | AJ719342.1 | - | 3 - 4 | 301 | 69 | |
More Information
Set Membership: |
|

STX7
Genomic Map