Assay Details
| Gene Symbol: VCPIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012948.1 | NP_001012966.1 | 2 - 3 | 3002 | 86 | |
| GenBank mRNA | AJ720863.1 | - | 2 - 3 | 3002 | 86 | |
More Information
Set Membership: |
|

VCPIP1
Genomic Map