Assay Details
| Gene Symbol: KDELR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012996.1 | NP_001013014.1 | 4 - 5 | 867 | 68 | |
| GenBank mRNA | BX935103.2 | - | 4 - 5 | 675 | 68 | |
More Information
Set Membership: |
|

KDELR3
Genomic Map