Assay Details
| Gene Symbol: ASS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013395.1 | NP_001013413.1 | 4 - 5 | 443 | 107 | |
| GenBank mRNA | AJ720611.1 | - | 4 - 5 | 443 | 107 | |
| BX930222.1 | - | 4 - 5 | 439 | 107 | ||
| CR407399.1 | - | 4 - 5 | 590 | 107 | ||
More Information
Set Membership: |
|

ASS1
Genomic Map