Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018012.1 | NP_001018012.1 | 4 - 5 | 865 | 62 | |
| XM_025154507.1 | XP_025010275.1 | 3 - 4 | 710 | 62 | ||
| GenBank mRNA | X02827.1 | - | 865 | 62 | ||
More Information
Set Membership: |
|

ALAS1
Genomic Map