Assay Details
| Gene Symbol: RBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024575.1 | NP_001019746.1 | 1 - 2 | 3045 | 143 | |
| GenBank mRNA | AY994153.1 | - | 1 - 2 | 3045 | 143 | |
More Information
Set Membership: |
|

RBP3
Genomic Map