Assay Details
| Gene Symbol: TNFSF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024578.1 | NP_001019749.1 | 3 - 4 | 316 | 64 | |
| GenBank mRNA | AB194710.1 | - | 3 - 4 | 316 | 64 | |
| AY954626.1 | - | 3 - 4 | 196 | 64 | ||
| BX930081.1 | - | 3 - 4 | 321 | 64 | ||
| CR353356.1 | - | 4 - 5 | 424 | 64 | ||
More Information
Set Membership: |
|

TNFSF15
Genomic Map