Assay Details
| Gene Symbol: CD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024582.1 | NP_001019753.1 | 5 - 6 | 1007 | 107 | |
| GenBank mRNA | AB204802.1 | - | 5 - 6 | 1054 | 107 | |
| AY375530.1 | - | 5 - 6 | 1007 | 107 | ||
| AY849320.1 | - | 5 - 6 | 1007 | 107 | ||
More Information
Set Membership: |
|

CD1B
Genomic Map