Assay Details
| Gene Symbol: PRLHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286221.2 | XP_015141707.1 | 2 | 7293 | 65 | |
| XM_015286222.2 | XP_015141708.1 | 2 | 7188 | 65 | ||
More Information
Set Membership: |
|

PRLHR2
Genomic Map