Assay Details
| Gene Symbol: TIRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024829.1 | NP_001020000.1 | 5 - 6 | 914 | 123 | |
| XM_015297978.2 | XP_015153464.1 | 6 - 7 | 1016 | 123 | ||
| XM_015297979.2 | XP_015153465.1 | 6 - 7 | 1067 | 123 | ||
| XM_015297983.2 | XP_015153469.1 | 5 - 6 | 1266 | 123 | ||
| XM_015297984.2 | XP_015153470.1 | 5 - 6 | 974 | 123 | ||
| XM_025143181.1 | XP_024998949.1 | 5 - 6 | 1649 | 123 | ||
| GenBank mRNA | BX929613.1 | - | 1 - 2 | 709 | 123 | |
| BX931654.2 | - | 5 - 6 | 917 | 123 | ||
| BX933959.1 | - | 6 - 7 | 1104 | 123 | ||
| CR390930.1 | - | 2 - 3 | 435 | 123 | ||
| DQ019929.1 | - | 5 - 6 | 914 | 123 | ||
More Information
Set Membership: |
|

TIRAP
Genomic Map