Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025304.1 | NP_001020475.1 | 2 - 3 | 725 | 76 | |
| XM_015296317.2 | XP_015151803.1 | 2 - 3 | 754 | 76 | ||
| XM_015296318.2 | XP_015151804.1 | 2 - 3 | 753 | 76 | ||
| XM_015296319.2 | XP_015151805.1 | 2 - 3 | 845 | 76 | ||
| XM_015296320.2 | XP_015151806.1 | 2 - 3 | 745 | 76 | ||
| GenBank mRNA | BX935980.1 | - | 1 - 2 | 567 | 76 | |
| U26645.1 | - | 2 - 3 | 725 | 76 | ||
More Information
Set Membership: |
|

BCL2L1
Genomic Map