Assay Details
| Gene Symbol: CHMP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025440.1 | NP_001020611.1 | 5 - 6 | 410 | 66 | |
| GenBank mRNA | BX950230.2 | - | 5 - 6 | 410 | 66 | |
| CR389488.1 | - | 1 - 2 | 112 | 66 | ||
| CR406221.1 | - | 5 - 6 | 406 | 66 | ||
More Information
Set Membership: |
|

CHMP1A
Genomic Map