Assay Details
| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030008.1 | NP_001025179.1 | 4 - 5 | 496 | 102 | |
| XM_015276902.2 | XP_015132388.1 | 4 - 5 | 2170 | 102 | ||
| XM_015276903.2 | XP_015132389.1 | 4 - 5 | 733 | 102 | ||
| XM_015276904.2 | XP_015132390.1 | 4 - 5 | 904 | 102 | ||
| XM_015276905.2 | XP_015132391.1 | 4 - 5 | 956 | 102 | ||
| GenBank mRNA | BX931554.1 | - | 4 - 5 | 484 | 102 | |
| CD762344.1 | - | 4 - 5 | 522 | 102 | ||
More Information
Set Membership: |
|

AP1S3
Genomic Map