Assay Details
| Gene Symbol: NATD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030633.1 | NP_001025804.1 | 2 - 3 | 355 | 62 | |
| GenBank mRNA | AJ720448.1 | - | 2 - 3 | 355 | 62 | |
More Information
Set Membership: |
|

NATD1
Genomic Map