Assay Details
| Gene Symbol: PGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030638.1 | NP_001025809.1 | 1 - 2 | 623 | 84 | |
| GenBank mRNA | AJ851389.1 | - | 1 - 2 | 623 | 84 | |
More Information
Set Membership: |
|

PGP
Genomic Map