Assay Details
| Gene Symbol: STK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030853.1 | NP_001026024.1 | 10 - 11 | 1375 | 62 | |
| XM_015296363.2 | XP_015151849.2 | 11 - 12 | 1461 | 62 | ||
| GenBank mRNA | AJ720430.1 | - | 10 - 11 | 1375 | 62 | |
More Information
Set Membership: |
|

STK4
Genomic Map