Assay Details
| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278151.1 | NP_001265080.1 | 11 - 12 | 2214 | 75 | |
| XM_015296380.2 | XP_015151866.1 | 11 - 12 | 2390 | 75 | ||
| GenBank mRNA | AJ719335.1 | - | 10 - 11 | 2104 | 75 | |
| BU243344.1 | - | 5 - 6 | 590 | 75 | ||
More Information
Set Membership: |
|

SLCO4A1
Genomic Map