Assay Details
| Gene Symbol: PRELID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030866.1 | NP_001026037.1 | 5 - 6 | 564 | 109 | |
| GenBank mRNA | AJ851573.1 | - | 5 - 6 | 564 | 109 | |
More Information
Set Membership: |
|

PRELID3B
Genomic Map