Assay Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030867.1 | NP_001026038.1 | 2 - 3 | 306 | 115 | |
| XM_015296407.1 | XP_015151893.1 | 2 - 3 | 309 | 115 | ||
| GenBank mRNA | AJ719816.1 | - | 2 - 3 | 306 | 115 | |
More Information
Set Membership: |
|

RAB22A
Genomic Map