Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030880.1 | NP_001026051.1 | 3 - 4 | 388 | 124 | |
| XM_015296896.1 | XP_015152382.1 | 5 - 6 | 2315 | 124 | ||
| XM_015296898.2 | XP_015152384.1 | 3 - 4 | 432 | 124 | ||
| XM_015296899.2 | XP_015152385.1 | 3 - 4 | 1356 | 124 | ||
| XM_015296900.2 | XP_015152386.1 | 3 - 4 | 1351 | 124 | ||
| XM_025142523.1 | XP_024998291.1 | 3 - 4 | 467 | 124 | ||
| XM_025142524.1 | XP_024998292.1 | 3 - 4 | 464 | 124 | ||
| XM_025142525.1 | XP_024998293.1 | 6 - 7 | 554 | 124 | ||
| XM_025142526.1 | XP_024998294.1 | 6 - 7 | 551 | 124 | ||
| GenBank mRNA | AJ720259.1 | - | 3 - 4 | 388 | 124 | |
More Information
Set Membership: |
|

SZRD1
Genomic Map