Assay Details
| Gene Symbol: SERINC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030890.1 | NP_001026061.1 | 1 - 2 | 75 | 51 | |
| GenBank mRNA | AJ720286.1 | - | 1 - 2 | 75 | 51 | |
More Information
Set Membership: |
|

SERINC2
Genomic Map