Assay Details
| Gene Symbol: OSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031077.1 | NP_001026248.1 | 5 - 6 | 894 | 63 | |
| GenBank mRNA | AJ719270.1 | - | 5 - 6 | 894 | 63 | |
More Information
Set Membership: |
|

OSTM1
Genomic Map