Assay Details
| Gene Symbol: MTMR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031101.1 | NP_001026272.1 | 2 - 3 | 425 | 68 | |
| XM_015285129.2 | XP_015140615.1 | 2 - 3 | 440 | 68 | ||
| GenBank mRNA | AJ851634.1 | - | 2 - 3 | 425 | 68 | |
More Information
Set Membership: |
|

MTMR9
Genomic Map