Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031102.1 | NP_001026273.1 | 9 - 10 | 1098 | 68 | |
| XM_015285137.2 | XP_015140623.1 | 9 - 10 | 928 | 68 | ||
| XM_015285138.2 | XP_015140624.1 | 9 - 10 | 820 | 68 | ||
| XM_015285139.2 | XP_015140625.1 | 9 - 10 | 822 | 68 | ||
| XM_015285140.2 | XP_015140626.1 | 9 - 10 | 925 | 68 | ||
| XM_025148804.1 | XP_025004572.1 | 10 - 11 | 1731 | 68 | ||
| GenBank mRNA | AJ720674.1 | - | 9 - 10 | 1098 | 68 | |
More Information
Set Membership: |
|

SUPT3H
Genomic Map