Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031181.2 | NP_001026352.2 | 2 - 3 | 936 | 63 | |
| XM_015286386.2 | XP_015141872.2 | 2 - 3 | 413 | 63 | ||
| XM_015286387.2 | XP_015141873.2 | 2 - 3 | 333 | 63 | ||
| XM_025150672.1 | XP_025006440.1 | 2 - 3 | 611 | 63 | ||
| GenBank mRNA | AJ851500.1 | - | 2 - 3 | 937 | 63 | |
More Information
Set Membership: |
|

WEE1
Genomic Map