Assay Details
| Gene Symbol: PDHX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031187.1 | NP_001026358.1 | 10 - 11 | 1260 | 95 | |
| XM_025150701.1 | XP_025006469.1 | 10 - 11 | 1141 | 95 | ||
| XM_025150702.1 | XP_025006470.1 | 10 - 11 | 1265 | 95 | ||
| GenBank mRNA | AJ851681.1 | - | 10 - 11 | 1260 | 95 | |
More Information
Set Membership: |
|

PDHX
Genomic Map