Assay Details
| Gene Symbol: KATNBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031198.1 | NP_001026369.1 | 4 - 5 | 518 | 78 | |
| XM_015287340.1 | XP_015142826.1 | 4 - 5 | 767 | 78 | ||
| XM_015287341.2 | XP_015142827.1 | 4 - 5 | 691 | 78 | ||
| GenBank mRNA | AJ720818.1 | - | 4 - 5 | 518 | 78 | |
More Information
Set Membership: |
|

KATNBL1
Genomic Map