Assay Details
| Gene Symbol: CCDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031221.1 | NP_001026392.1 | 4 - 5 | 762 | 71 | |
| GenBank mRNA | AJ851457.1 | - | 4 - 5 | 762 | 71 | |
More Information
Set Membership: |
|

CCDC6
Genomic Map