Assay Details
| Gene Symbol: GNAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031427.1 | NP_001026598.1 | 3 - 4 | 631 | 88 | |
| XM_025144719.1 | XP_025000487.1 | 3 - 4 | 368 | 88 | ||
| GenBank mRNA | AJ851735.1 | - | 3 - 4 | 631 | 88 | |
More Information
Set Membership: |
|

GNAQ
Genomic Map