Assay Details
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204380.2 | NP_989711.2 | 4 - 5 | 509 | 75 | |
GenBank mRNA | AF459286.1 | - | 4 - 5 | 518 | 75 | |
AF521908.1 | - | 4 - 5 | 518 | 75 | ||
AJ851624.1 | - | 4 - 5 | 509 | 75 | ||
AY237359.1 | - | 4 - 5 | 512 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
